Fmoc-L-homopropargylglycine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N,Fmoc-L-homopropargylglycine, Fmoc-HoGly(Propargyl)-OH, (2S)-2-(Fmoc-amino)-5-hexynoic acid
BICR102-500MG
APOSBICR102-500MGEA
1420
EUR
APOSBICR102-500MG
Fmoc-L-homopropargylglycine
Fmoc-L-homopropargylglycine
Formel:
C₂₁H₁₉NO₄ Molecular Weight: 349,38 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD11052899 CAS-Nummer: 942518-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...