(2S,5R)-5-hydroxypipecolic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S,5R)-5-hydroxypiperidine-2-carboxylic acid hydrochloride

BICR111-100MG
APOSBICR111-100MGEA 343 EUR
APOSBICR111-100MG
(2S,5R)-5-hydroxypipecolic acid hydrochloride
(2S,5R)-5-hydroxypipecolic acid hydrochloride
Formel: C₆H₁₁NO₃·ClH
Molecular Weight: 181,617 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07776737
CAS-Nummer: 824943-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR