Nijmegen-1

Lieferant: Apollo Scientific

Synonyme: (Z)-methyl 2-(1,3-dioxoisoindolin-2-yl)-3-((4-methyl-5-oxo-2,5-dihydrofuran-2-yl)oxy)acrylate

BICR134-5MG
APOSBICR134-5MGEA 429 EUR
APOSBICR134-5MG
Nijmegen-1
Nijmegen-1
Formel: C₁₇H₁₃NO₇
Molecular Weight: 343,288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24448757
CAS-Nummer: 159155-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR