Mono-6-O-(p-toluenesulphonyl)-β-cyclodextrin

Lieferant: Apollo Scientific

Synonyme: Mono-(6-p-toluenesulphonyl)-β-cyclodextrin

BICY1015-5G
APOSBICY1015-5GEA 401 EUR
APOSBICY1015-5G
Mono-6-O-(p-toluenesulphonyl)-β-cyclodextrin
Mono-6-O-(p-toluenesulphonyl)-beta-cyclodextrin
Formel: C₄₉H₇₆O₃₇S
Molecular Weight: 1289,1706 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864975
CAS-Nummer: 67217-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR