Trimethyl-β-cyclodextrin

Lieferant: Apollo Scientific

BICY1077-5G
APOSBICY1077-5GEA 383 EUR
APOSBICY1077-5G
Trimethyl-β-cyclodextrin
Trimethyl-beta-cyclodextrin
Formel: C₆₃H₁₁₂O₃₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00010728
CAS-Nummer: 55216-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR