6A-Azido-6A-deoxy-beta-cyclodextrin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Mono(6-azido-6-desoxy)-β-cyclodextrin, Mono-6A-azido-β-cyclodextrin, 6-Deoxy-6-azido-β-cyclodextrin, 6-Monoazido-6-monodeoxy-β-cyclodextrin, 6-Azido-6-deoxycyclomaltoheptaose, 6-Monoazido-6-deoxy-β-cyclodextrin, 6-Monodeoxy-6-monoazido-beta-cyclodextrin, 2,4,7,9,12,14,17,19,22,24,27,29,32,34-Tetradecaoxaoctacyclo[31.2.2.23,6.28,11.213,16.218,21.223,26.228,31]nonatetracontane, β-cyclodextrin derivative, 6-Monoazido-β-cyclodextrin, 6A-Azido-6A-deoxy-β-cyclodextrin
BICY1079-1G
APOSBICY1079-1GEA
719
EUR
APOSBICY1079-1G
6A-Azido-6A-deoxy-beta-cyclodextrin
6A-Azido-6A-deoxy-beta-cyclodextrin
Formel:
C₄₂H₆₉N₃O₃₄ Molecular Weight: 1160,00 g/mol Schmelzpunkt: 206 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864973 CAS-Nummer: 98169-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...