4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) 92%

Lieferant: Apollo Scientific
Achtung

BID0433-100MG BID0433-250MG BID0433-1G
APOSBID0433-100MGEA 0 EUR
APOSBID0433-100MG APOSBID0433-250MG APOSBID0433-1G
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) 92%
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)

Dye used in the staining of DNA.

Formel: C₁₆H₁₅N₅·2HCl
Lagertemperatur: Kühlschrank
MDL: MFCD00012681
CAS-Nummer: 28718-90-3
EINECS: 249-186-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR