Dextransulfat Natriumsalz from leuconostoc 500,000
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dextran sulphate sodium from Dextran
BID1342-10G
APOSBID1342-10GEA
72.5
EUR
APOSBID1342-10G
Dextransulfat Natriumsalz from leuconostoc 500,000
Dextran sulphate sodium salt
Produced from Dextran 50,000.
- Free sulphate <0.2%
- Tested for suitability in nucleic acid hybridisations
Formel:
(C₆H₇Na₃O₁₄S₃)n Lagertemperatur: Kühlschrank |
MDL:
MFCD00081551 CAS-Nummer: 9011-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...