(3,4-Dihydroxyphenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid

BID2105-100G
APOSBID2105-100GEA 142 EUR
APOSBID2105-100G
(3,4-Dihydroxyphenyl)essigsäure 98%
(3,4-Dihydroxyphenyl)essigsäure
Formel: (C₆H₃(OH)₂)CH₂COOH
Lagertemperatur: Raumtemperatur
MDL: MFCD00004338
CAS-Nummer: 102-32-9
EINECS: 203-024-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR