5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)

Lieferant: Apollo Scientific
Achtung

BID2180-1G BID2180-25G BID2180-5G
APOSBID2180-1GEA 21.9 EUR
APOSBID2180-1G APOSBID2180-25G APOSBID2180-5G
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)

Used for measurement of sulphhydryl content inpeptides, proteins and tissue.

Formel: C₁₄H₈N₂O₈S₂
Schmelzpunkt: 233…243 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007140
CAS-Nummer: 69-78-3
EINECS: 200-714-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR