Decyl-β-D-glucopyranosid

Lieferant: Apollo Scientific
Achtung

BID6001-1G BID6001-5G BID6001-10G
APOSBID6001-1GEA 95.7 EUR
APOSBID6001-1G APOSBID6001-5G APOSBID6001-10G
Decyl-β-D-glucopyranosid
Decyl-β-D-glucopyranosid
Formel: C₁₆H₃₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00063297
CAS-Nummer: 58846-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR