Marein with hplc

Lieferant: Apollo Scientific
Achtung

Synonyme: (E)-1-(2,3-Dihydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)phenyl)-3-(3,4-dihydroxyphenyl)prop-2-en-1-one, Okanin-4'-O-glucoside hplc, Oglucoside

BIDF1016-5MG
APOSBIDF1016-5MGEA 197 EUR
APOSBIDF1016-5MG
Marein with hplc
Marein
Formel: C₂₁H₂₂O₁₁
Molecular Weight: 450,393 g/mol
Schmelzpunkt: 197…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017496
CAS-Nummer: 535-96-6
EINECS: 208-623-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR