Gossypin

Lieferant: Apollo Scientific

BIDF1030-250MG BIDF1030-50MG BIDF1030-100MG
APOSBIDF1030-250MGEA 261 EUR
APOSBIDF1030-250MG APOSBIDF1030-50MG APOSBIDF1030-100MG
Gossypin
Gossypin
Lagertemperatur: Raumtemperatur
CAS-Nummer: 652-78-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR