Piperine 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2E,4E)-5-(Benzo[d][1,3]dioxol-5-yl)-1-(piperidin-1-yl)penta-2,4-dien-1-one, 1-[5-(1,3-Benzodioxol-5-yl)-2,4-pentadienoyl]piperidine, 1-Piperoylpiperidine, 1-Piperylpiperidine

BIDF1038-25G BIDF1038-5G
APOSBIDF1038-25GEA 114 EUR
APOSBIDF1038-25G APOSBIDF1038-5G
Piperine 96%
Piperine

Natural product.

Formel: C₁₇H₁₉NO₃
Molecular Weight: 285.34 g/mol
Schmelzpunkt: 131…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005839
CAS-Nummer: 94-62-2
EINECS: 202-348-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR