Epiamastatin hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,3R)-AHMHA-Val-Val-Asp-OH HCl

BIE0143-5MG
APOSBIE0143-5MGEA 509 EUR
APOSBIE0143-5MG
Epiamastatin hydrochloride
Epiamastatin hydrochloride

Diastereoepimer of aminopeptidase inhibitor amastatin

Formel: C₂₁H₃₈N₄O₈·ClH
Molecular Weight: 511,009 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00070396
CAS-Nummer: 100992-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR