24-Epibrassinolide
Lieferant: Apollo Scientific
|
Achtung
|
BIE0244-100MG
BIE0244-10MG
APOSBIE0244-100MGEA
0
EUR
APOSBIE0244-100MG
APOSBIE0244-10MG
24-Epibrassinolide
24-Epibrassinolide
Formel:
C₂₈H₄₈O₆ Schmelzpunkt: 254…256 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00133310 CAS-Nummer: 78821-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...