Fumitremorgin C

Lieferant: Apollo Scientific
Achtung

BIF1010-10MG BIF1010-1MG BIF1010-5MG
APOSBIF1010-10MGEA 1900 EUR
APOSBIF1010-10MG APOSBIF1010-1MG APOSBIF1010-5MG
Fumitremorgin C
Fumitremorgin C
Formel: C₂₂H₂₅N₃O₃
Lagertemperatur: Kühlschrank
CAS-Nummer: 118974-02-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR