Riboflavin-5'-phosphat Natriumsalz USP
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Vitamin B2 phosphate sodium salt, Riboflavin 5'-Monophosphate sodium salt, Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogen phosphate, Flavin mononucleotide, Lactoflavin phosphate sodium salt, Riboflavin phosphate sodium
BIF4012-100G
BIF4012-25G
BIF4012-1KG
APOSBIF4012-100GEA
509
EUR
APOSBIF4012-100G
APOSBIF4012-25G
APOSBIF4012-1KG
Riboflavin-5'-phosphat Natriumsalz USP
Riboflavin-5'-phosphat Natriumsalz
Formel:
C₁₇H₂₀N₄NaO₉P Molecular Weight: 478.3256 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00065362 CAS-Nummer: 130-40-5 EINECS: 204-988-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...