Riboflavin-5'-phosphat Natriumsalz USP

Lieferant: Apollo Scientific
Achtung

Synonyme: Vitamin B2 phosphate sodium salt, Riboflavin 5'-Monophosphate sodium salt, Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogen phosphate, Flavin mononucleotide, Lactoflavin phosphate sodium salt, Riboflavin phosphate sodium

BIF4012-100G BIF4012-25G BIF4012-1KG
APOSBIF4012-100GEA 509 EUR
APOSBIF4012-100G APOSBIF4012-25G APOSBIF4012-1KG
Riboflavin-5'-phosphat Natriumsalz USP
Riboflavin-5'-phosphat Natriumsalz
Formel: C₁₇H₂₀N₄NaO₉P
Molecular Weight: 478.3256 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00065362
CAS-Nummer: 130-40-5
EINECS: 204-988-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR