Quizartinib 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: AC220, 1-(5-(tert-Butyl)isoxazol-3-yl)-3-(4-(7-(2-morpholinoethoxy)benzo[d]imidazo[2,1-b]thiazol-2-yl)phenyl)urea

BIFK0001-250MG
APOSBIFK0001-250MGEA 154 EUR
APOSBIFK0001-250MG
Quizartinib 99%
Quizartinib

  • Free Base, >99%

Formel: C₂₉H₃₂N₆O₄S
Molecular Weight: 560,7 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD18074524
CAS-Nummer: 950769-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR