Topotecan hydrochloride

Lieferant: Apollo Scientific

BIFK0005-100MG
APOSBIFK0005-100MGEA 21.5 EUR
APOSBIFK0005-100MG
Topotecan hydrochloride
Topotecan hydrochloride
Formel: C₂₃H₂₃N₃O₅.HCl
Lagertemperatur: Gefrierschrank
MDL: MFCD00866235
CAS-Nummer: 119413-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR