Vinorelbine tartrate

Lieferant: Apollo Scientific

Synonyme: Methyl (3aR,3a1R,4R,5S,5aR,10bR)-4-acetoxy-3a-ethyl-9-((8R)-4-ethyl-8-(methoxycarbonyl)-1,3,6,7,8,9-hexahydro-2,6-methanoazecino[4,3-b]indol-8-yl)-5-hydroxy-8-methoxy-6-methyl-3a,3a1,4,5,5a,6,11,12-octahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylate bis((2R,3R)-2,3-dihydroxysuccinate), Vinorelbine ditartrate

BIFK0008-10MG BIFK0008-25MG BIFK0008-50MG BIFK0008-1G
APOSBIFK0008-10MGEA 109 EUR
APOSBIFK0008-10MG APOSBIFK0008-25MG APOSBIFK0008-50MG APOSBIFK0008-1G
Vinorelbine tartrate
Vinorelbine tartrate

Ditartrate Salt, >99%

Formel: C₅₃H₆₆N₄O₂₀
Molecular Weight: 1079.106 g/mol
Schmelzpunkt: 167…180 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD03613607
CAS-Nummer: 125317-39-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR