CUDC-101 freie Base ≥99%

Lieferant: Apollo Scientific

BIFK0015-10MG BIFK0015-5MG BIFK0015-1MG
APOSBIFK0015-10MGEA 0 EUR
APOSBIFK0015-10MG APOSBIFK0015-5MG APOSBIFK0015-1MG
CUDC-101 freie Base ≥99%
CUDC-101
Formel: C₂₄H₂₆N₄O₄
Lagertemperatur: Gefrierschrank
MDL: MFCD15528940
CAS-Nummer: 1012054-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR