Imatinib ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide
BIFK0025-10G
BIFK0025-1G
BIFK0025-5G
APOSBIFK0025-10GEA
248
EUR
APOSBIFK0025-10G
APOSBIFK0025-1G
APOSBIFK0025-5G
Imatinib ≥99%
Imatinib
Imatinib is the active ingredient in the drug sold under the trade name Gleevec® in USA or Glivec in Europe and Australia.
Formel:
C₂₉H₃₁N₇O Molecular Weight: 493.62 g/mol Schmelzpunkt: 209…221 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD05662257 CAS-Nummer: 152459-95-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...