Imatinib ≥99%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide

BIFK0025-10G BIFK0025-1G BIFK0025-5G
APOSBIFK0025-10GEA 248 EUR
APOSBIFK0025-10G APOSBIFK0025-1G APOSBIFK0025-5G
Imatinib ≥99%
Imatinib

Imatinib is the active ingredient in the drug sold under the trade name Gleevec® in USA or Glivec in Europe and Australia.

Formel: C₂₉H₃₁N₇O
Molecular Weight: 493.62 g/mol
Schmelzpunkt: 209…221 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD05662257
CAS-Nummer: 152459-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR