Imatinib mesylate ≥99%
Lieferant: Apollo Scientific
Synonyme:
N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide methanesulphonate, Imatinib Methanesulphonate
BIFK0026-1G
BIFK0026-5G
BIFK0026-250MG
APOSBIFK0026-1GEA
116
EUR
APOSBIFK0026-1G
APOSBIFK0026-5G
APOSBIFK0026-250MG
Imatinib mesylate ≥99%
Imatinib mesylate
Imatinib is the active ingredient in the drug sold under the trade name Gleevec® in USA or Glivec in Europe and Australia.
Formel:
C₂₉H₃₁N₇O·CH₄SO₃ Molecular Weight: 589.72 g/mol Schmelzpunkt: 203…224 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD04307699 CAS-Nummer: 220127-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...