Motesanib diphosphate

Lieferant: Apollo Scientific

Synonyme: N-(3,3-Dimethylindolin-6-yl)-2-((pyridin-4-ylmethyl)amino)nicotinamide diphosphate, N-(3,3-dimethyl-1,2-dihydroindol-6-yl)-2-(pyridin-4-ylmethylamino)pyridine-3-carboxamide phosphoric acid

BIFK0030-10MG BIFK0030-50MG BIFK0030-100MG
APOSBIFK0030-10MGEA 65.4 EUR
APOSBIFK0030-10MG APOSBIFK0030-50MG APOSBIFK0030-100MG
Motesanib diphosphate
Motesanib diphosphate

Diphosphate Salt, >99%

Formel: C₂₂H₂₉N₅O₉P₂
Molecular Weight: 569.4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD12407403
CAS-Nummer: 857876-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR