NVP-AUY922 freie Base ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-[2,4-Dihydroxy-5-isopropylphenyl]-N-ethyl-4-{4-[(morpholin-4-yl)methyl]phenyl}isoxazole-3-carboxam, Luminespib, 5-(2,4-Dihydroxy-5-isopropylphenyl)-N-ethyl-4-(4-(morpholinomethyl)phenyl)isoxazole-3-carboxamide
BIFK0032-5MG
APOSBIFK0032-5MGEA
27.7
EUR
APOSBIFK0032-5MG
NVP-AUY922 freie Base ≥99%
NVP-AUY922
Formel:
C₂₆H₃₁N₃O₅ Molecular Weight: 465,54 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD14635361 CAS-Nummer: 747412-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...