Clofarabin freie Base

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, (2R,3R,4S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)tetrahydrofuran-3-ol, 2-Chloro-9-(2-deoxy-2-fluoro-beta-D-arabinofuranosyl)adenine

BIFK0045-1G
APOSBIFK0045-1GEA 256 EUR
APOSBIFK0045-1G
Clofarabin freie Base
Clofarabine

Clofarabine is a second generation purine nucleoside analogue with multiple properties such as inhibition of ribonucleotide reductase, incorporation into DNA, and induction of apoptosis.

Formel: C₁₀H₁₁ClFN₅O₃
Schmelzpunkt: 230…240 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00871077
CAS-Nummer: 123318-82-1
UN: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR