5-Aza-2'-deoxycytidine freie Base
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one
BIFK0051-500MG
BIFK0051-50MG
BIFK0051-100MG
APOSBIFK0051-500MGEA
400
EUR
APOSBIFK0051-500MG
APOSBIFK0051-50MG
APOSBIFK0051-100MG
5-Aza-2'-deoxycytidine freie Base
5-Aza-2'-deoxycytidine
- Purity >99%
Formel:
C₈H₁₂N₄O₄ Molecular Weight: 228.21 g/mol Schmelzpunkt: 193…203 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00043011 CAS-Nummer: 2353-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...