Genistin

Lieferant: Apollo Scientific

Synonyme: 5-Hydroxy-3-(4-hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

BIG0101-100MG BIG0101-25MG
APOSBIG0101-100MGEA 422 EUR
APOSBIG0101-100MG APOSBIG0101-25MG
Genistin
Genistin

Inactive analog of genistein, useful as a negativecontrol for genistein and other tyrosine inhibitors

Formel: C₂₁H₂₀O₁₀
Lagertemperatur: Gefrierschrank
MDL: MFCD00016883
CAS-Nummer: 529-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR