G418 Disulfat

Lieferant: Apollo Scientific
Gefahr

Synonyme: G-418 Sulfat, G418 Sulfat, Geneticin®, G-418 Disulfat

BIG0175-1G BIG0175-5G
APOSBIG0175-1GEA 31.9 EUR
APOSBIG0175-1G APOSBIG0175-5G
G418 Disulfat
G418 disulphate

Aminoglycoside antibiotic.

Formel: C₂₀H₄₄N₄O₁₈S₂
Lagertemperatur: Kühlschrank
MDL: MFCD00058314
CAS-Nummer: 108321-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR