D-Glucoronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: D-(+)Glucuronic acid, D-Glucuronic acid, D-(+)-glucuronic acid, (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid

BIG1006-100G BIG1006-25G BIG1006-50G
APOSBIG1006-100GEA 299 EUR
APOSBIG1006-100G APOSBIG1006-25G APOSBIG1006-50G
D-Glucoronic acid
D-Glucoronic acid
Formel: C₆H₁₀O₇
Molecular Weight: 194.139 g/mol
Schmelzpunkt: 158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00166981
CAS-Nummer: 6556-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR