D-Glucoronic acid

Lieferant: Apollo Scientific
Achtung

BIG1006-100G BIG1006-25G BIG1006-50G
APOSBIG1006-100GEA 299 EUR
APOSBIG1006-100G APOSBIG1006-25G APOSBIG1006-50G
D-Glucoronic acid
D-Glucoronic acid
Formel: C₆H₁₀O₇
Schmelzpunkt: 158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00166981
CAS-Nummer: 6556-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR