L-Glucono-1,5-lactone ≥85%

Lieferant: Apollo Scientific

Synonyme: (3S,4R,5R,6S)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one

BIG1121-100MG BIG1121-1G BIG1121-250MG
APOSBIG1121-100MGEA 241 EUR
APOSBIG1121-100MG APOSBIG1121-1G APOSBIG1121-250MG
L-Glucono-1,5-lactone ≥85%
L-Glucono-1,5-lactone
Formel: C₆H₁₀O₆
Schmelzpunkt: 135 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00067388
CAS-Nummer: 52153-09-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR