D(+)-Galactose
Lieferant: Apollo Scientific
Synonyme:
(2R,3S,4S,5R)-Pentahydroxyhexanal
BIG1724-1KG
BIG1724-500G
BIG1724-5KG
APOSBIG1724-1KGEA
109
EUR
APOSBIG1724-1KG
APOSBIG1724-500G
APOSBIG1724-5KG
D(+)-Galactose
D(+)-Galactose
Formel:
C₆H₁₂O₆ Molecular Weight: 180.16 g/mol Siedepunkt: 527 Schmelzpunkt: 168…170 °C Dichte: 1.5 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151230 CAS-Nummer: 59-23-4 EINECS: 200-416-4 Merck Index: 13,04356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...