Guanosin-5'-triphosphat Dinatriumsalz
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Guanosine-5'-triphosphate disodium salt, GTP disodium salt, Guanosine-5-triphosphoric aicd disodium salt
BIG4005-1G
BIG4005-250MG
BIG4005-500MG
APOSBIG4005-1GEA
109
EUR
APOSBIG4005-1G
APOSBIG4005-250MG
APOSBIG4005-500MG
Guanosin-5'-triphosphat Dinatriumsalz
Guanosin-5'-triphosphat Dinatriumsalz
Formel:
C₁₀H₁₄N₅Na₂O₁₄P₃ Lagertemperatur: Gefrierschrank |
MDL:
MFCD03410297 CAS-Nummer: 56001-37-7 EINECS: 259-940-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...