Hydroxyharnstoff 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Hydroxycarbamid, Aminoameisensäurehydroxyamid

BIH0153-10G BIH0153-25G BIH0153-5G
APOSBIH0153-10GEA 43.5 EUR
APOSBIH0153-10G APOSBIH0153-25G APOSBIH0153-5G
Hydroxyharnstoff 98%
Hydroxyharnstoff

Anti-neoplastic. Inactivates ribonucleoside reductase byformation of free radical nitroxide. This blocks synthesis of deoxynucleotides which inhibits DNA synthesis.


  • Protein

Formel: CH₄N₂O₂
Molecular Weight: 76.05 g/mol
Siedepunkt: 296
Schmelzpunkt: 135 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00007943
CAS-Nummer: 127-07-1
EINECS: 204-821-7
Merck Index: 13,04874

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR