≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulphonic acid), HEPPS, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulphonic acid, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulphonic acid, 4-(2-Hydroxyethyl)piperazine-1-propanesulphonic acid
BIH0601-250G
BIH0601-100G
BIH0601-500G
APOSBIH0601-250GEA
174
EUR
APOSBIH0601-250G
APOSBIH0601-100G
APOSBIH0601-500G
≥99%
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinpropansulfonsäure)
Biological buffer - useful range 7.3-8.7
Formel:
C₉H₂₀N₂O₄S Schmelzpunkt: 237…239 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006160 CAS-Nummer: 16052-06-5 EINECS: 240-198-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...