HEPPSO (β-Hydroxy-4-(2-hydroxyethyl)piperazin-1-propansulfonsäure) ≥98%
Lieferant: Apollo Scientific
Synonyme:
3-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-hydroxypropanesulphonic acid
BIH0893-100G
APOSBIH0893-100GEA
480
EUR
APOSBIH0893-100G
HEPPSO (β-Hydroxy-4-(2-hydroxyethyl)piperazin-1-propansulfonsäure) ≥98%
HEPPSO (β-Hydroxy-4-(2-hydroxyethyl)piperazin-1-propansulfonsäure)
Biological buffer - useful range 7.1-8.5
Formel:
C₉H₂₀N₂O₅S Molecular Weight: 268.3305 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038351 CAS-Nummer: 68399-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...