Fasudil dihydrochloride ≥99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(5-Isoquinolinesulphonyl)homopiperazine dihydrochloride salt, HA-1077 dihydrochloride salt

BIH1037-10MG
APOSBIH1037-10MGEA 435 EUR
APOSBIH1037-10MG
Fasudil dihydrochloride ≥99%
Fasudil dihydrochloride

Kinase inhibitor

Formel: C₁₄H₁₇N₃O₂S·2HCl
Molecular Weight: 364,29 g/mol
Schmelzpunkt: 248…249 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00153805
CAS-Nummer: 203911-27-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR