DL-Homocysteinthiolactonhydrochlorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
DL-3-Aminotetrahydrothiophen-2-one Hydrochloride, 3-Aminodihydrothiophen-2(3H)-one hydrochloride, DL-Homocysteine thiolactone HCl
BIH4018-100G
BIH4018-25G
APOSBIH4018-100GEA
69.6
EUR
APOSBIH4018-100G
APOSBIH4018-25G
DL-Homocysteinthiolactonhydrochlorid 97%
DL-Homocysteinethiolactone hydrochloride
Formel:
C₄H₇NOS Schmelzpunkt: 202…204 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012724 CAS-Nummer: 6038-19-3 EINECS: 227-923-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...