DL-Homocysteinthiolactonhydrochlorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: DL-3-Aminotetrahydrothiophen-2-one Hydrochloride, 3-Aminodihydrothiophen-2(3H)-one hydrochloride, DL-Homocysteine thiolactone HCl

BIH4018-100G BIH4018-25G
APOSBIH4018-100GEA 69.6 EUR
APOSBIH4018-100G APOSBIH4018-25G
DL-Homocysteinthiolactonhydrochlorid 97%
DL-Homocysteinethiolactone hydrochloride
Formel: C₄H₇NOS
Schmelzpunkt: 202…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012724
CAS-Nummer: 6038-19-3
EINECS: 227-923-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR