N-Hydroxysuccinimid ≥97%

Lieferant: Apollo Scientific
Achtung

BIH6094-1KG BIH6094-250G BIH6094-100G
APOSBIH6094-1KGEA 122 EUR
APOSBIH6094-1KG APOSBIH6094-250G APOSBIH6094-100G
N-Hydroxysuccinimid ≥97%
N-Hydroxysuccinimid
Formel: C₄H₅NO₃
Schmelzpunkt: 94…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005516
CAS-Nummer: 6066-82-6
EINECS: 228-001-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR