1,3,4,6-Tetrachloro-3-6-diphenylglycouril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Iodo-Gen®, NSC 4462
BII100-1G
APOSBII100-1GEA
255
EUR
APOSBII100-1G
1,3,4,6-Tetrachloro-3-6-diphenylglycouril
1,3,4,6-Tetrachloro-3-6-diphenylglycouril
Iodination reagent
Ref: Fraker,P.J. and Speck, J.C. (1978) Biophys.Comm. 80, 849-857
Formel:
C₁₆H₁₀Cl₄N₄O₂ Molecular Weight: 432,088 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00027367 CAS-Nummer: 51592-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...