3-Maleimidophenyl boronic acid
Lieferant: Apollo Scientific
BII108-100MG
APOSBII108-100MGEA 509 EUR
APOSBII108-100MG
3-Maleimidophenyl boronic acid
3-Maleimidophenyl boronic acid
Sulphhydryl reactive -Iodinatable