Candida antarctica Lipase A
Lieferant: Apollo Scientific
BIIM1000-100G BIIM1000-25G
APOSBIIM1000-100GEA 0 EUR
APOSBIIM1000-100G APOSBIIM1000-25G
Candida antarctica Lipase A
Enzyme
Covalent on IB-150A. Activity: 2000 TBU/g
  • immobilized on Immobead 150