Candida rugosa Lipase
Lieferant: Apollo Scientific
BIIM1004-100G BIIM1004-10G BIIM1004-25G
APOSBIIM1004-100GEA 0 EUR
APOSBIIM1004-100G APOSBIIM1004-10G APOSBIIM1004-25G
Candida rugosa Lipase
Enzyme
Covalent on IB-150A. Activity: 250 TBU/g
  • immobilized on Immobead 150