Candida rugosa Lipase
Lieferant: Apollo Scientific
BIIM1004-100G
BIIM1004-10G
BIIM1004-25G
APOSBIIM1004-100GEA
0
EUR
APOSBIIM1004-100G
APOSBIIM1004-10G
APOSBIIM1004-25G
Candida rugosa Lipase
Enzyme
Covalent on IB-150A. Activity: 250 TBU/g
- immobilized on Immobead 150