Sodium-2-oxobutyrate 97+%
Lieferant: Apollo Scientific
|
Achtung
|
BIK8008-1G
BIK8008-5G
APOSBIK8008-1GEA
0
EUR
APOSBIK8008-1G
APOSBIK8008-5G
Sodium-2-oxobutyrate 97+%
Sodium-2-oxobutyrate
Formel:
C₄H₆O₃ Schmelzpunkt: 210 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00064194 CAS-Nummer: 2013-26-5 EINECS: 217-937-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...