D(+)-Maltose Monohydrat

Lieferant: Apollo Scientific
Achtung

BIK9000-1KG BIK9000-500G
APOSBIK9000-1KGEA 79.9 EUR
APOSBIK9000-1KG APOSBIK9000-500G
D(+)-Maltose Monohydrat
D(+)-Maltose Monohydrat
Formel: C₁₂H₂₂O₁₁·1H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00149343
CAS-Nummer: 6363-53-7
EINECS: 200-716-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR